Learn More
Bachem Abz-Gly-p-nitro-Phe-Pro-OH
SDP
Supplier:  Bachem 4003531.0050
Encompass_Preferred
Abz-Gly-p-nitro-Phe-Pro-OH Trifluoroacetate, 67482-93-3, C23H25N5O7, Mr 483.48, 50mg. Fluorogenic (FRET) substrate for angiotensin I-converting enzyme (ACE). Enzymatic cleavage of Abz-Gly-Nph-Pro yields Abz-Gly-OH emitting at 420 nm.
Catalog No. 50-260-445