Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abz-Gly-p-nitro-Phe-Pro-OH | 67482-93-3 | 99.9% | 483.47 | 25 MG
SDP

Catalog No. 5000400266
Encompass_Preferred

Abz-Gly-p-nitro-Phe-Pro-OH is a fluorescent substrate for angiotensin I converting enzyme (ACE-I). It features excitation at 355 nm and emission at 405 nm, making it suitable for various research applications.

  • Fluorescent substrate for ACE-I
  • Excitation wavelength: 355 nm
  • Emission wavelength: 405 nm
  • Off-white to light yellow solid appearance
  • Purity: 99.9%
  • Molecular weight: 483.47
  • For research use only

Catalog No. 50-004-00266 Supplier Medchemexpress LLC Supplier No. 25MGHYP4545
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abz-Gly-p-nitro-Phe-Pro-OH is a fluorescent substrate for angiotensin I converting enzyme (ACE-I). It features excitation at 355 nm and emission at 405 nm, making it suitable for various research applications.

  • Fluorescent substrate for ACE-I
  • Excitation wavelength: 355 nm
  • Emission wavelength: 405 nm
  • Off-white to light yellow solid appearance
  • Purity: 99.9%
  • Molecular weight: 483.47
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.