Learn More
Carbosynth LLC. Abz-Leu-Ala-Gln-Ala-Val-Arg-Ser-Ser-Ser-Arg-Dap(Dnp)-NH2, 1mg, MW: 1444.54, Molecular formula: C59H93N23O20, CAS# 158442-41-2
SDP
Supplier:  Carbosynth LLC. SDP3818PI1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: 2-Aminobenzoyl-L-Leucyl-L-Alanyl-L-Glutaminyl-L-Alanyl-L-Valyl-L-Arginyl-L-Seryl-L-Seryl-L-Seryl-L-Arginyl-[N-- (2,4-Dinitrophenyl)-L-2,3-Diaminopropionyl] Amide, Application: Fluorescence-Quenching Substrate for ADAM17 / TNF- Converting Enzyme (TACE), Storage Temperature: -20C, References: G. Jin, X. Huang, R. Black, M. Wolfson, C. Rauch, H. McGregor, G. Ellestad, R. Cowling, Anal. Biochem., 302, 269 (2002).
Catalog No. 50-168-7052