Learn More
Carbosynth LLC. Abz-Ser-Pro-Tyr(NO2)-OH, 5mg, MW: (M.W. 529.51), Molecular formula: C24H27N5O9
SDP
Supplier:  Carbosynth LLC. SFQ3819PI5MG
Encompass_Preferred
Application: Fluorescence-Quenching Substrate for ACE2 (TBC5182), Storage Temperature: -20°C, References: Z.-H. Yan, K.-J. Ren, Y. Wang, S. Chen, T.A. Brock, and A. Rege, Anal. Biochem., 312, 141 (2003).
Catalog No. 50-168-7085