Learn More
Carbosynth LLC. Abz-Val-Ala-Asp-Nva-Arg-Asp-Arg-Gln-EDDnp, 1mg, MW: 1285.3, Molecular formula: C53H80N20O18, CAS# 1042405-14-0
SDP
Supplier:  Carbosynth LLC. SNP3232V1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: 2-Aminobenzoyl-L-valyl-L-alanyl-L-aspartyl-L-norvalyl-L-arginyl-L-aspartyl-L-arginyl-L-glutamine 2-(2,4-dinitrophenyl)aminoethyl-amide, Abz-VADnVRDRQ-EDDnp, nV = Nva, Application: Fluorescence-Quenching Substrate for Human Neutrophil Proteinase 3, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20C, References: B. Korkmaz, et al., J. Biol. Chem., 282, 1989 (2007)., B. Korkmaz, et al., Nat. Protoc., 3, 991 (2008).
Catalog No. 50-168-7140