Learn More
Medchemexpress LLC Abz-Val-Ala-Asp-Nva-Arg-Asp-Arg-Gln-EDDnp | PR3 substrate | 1042405-14-0 | 99.5% | 1285.33 | 10 MG
SDP
Catalog No. 5000423217
Encompass_Preferred
Abz-Val-Ala-Asp-Nva-Arg-Asp-Arg-Gln-EDDnp is a fluorescence-quenched peptide substrate for human proteinase 3 (PR3), demonstrating high catalytic efficiency (kcat/Km = 1,570 mM-1s-1). This substrate is ideal for detecting PR3 activity in various research applications, offering a sensitive and specific tool for studying proteolytic pathways.
  • Fluorescence-quenched peptide substrate
  • Specifically designed for human proteinase 3
  • Exhibits high catalytic efficiency
  • Enables detection of proteinase 3 activity
Catalog No. 50-004-23217 Supplier Medchemexpress LLC Supplier No. HYP1046110MG
Abz-Val-Ala-Asp-Nva-Arg-Asp-Arg-Gln-EDDnp is a fluorescence-quenched peptide substrate for human proteinase 3 (PR3), demonstrating high catalytic efficiency (kcat/Km = 1,570 mM-1s-1). This substrate is ideal for detecting PR3 activity in various research applications, offering a sensitive and specific tool for studying proteolytic pathways.
  • Fluorescence-quenched peptide substrate
  • Specifically designed for human proteinase 3
  • Exhibits high catalytic efficiency
  • Enables detection of proteinase 3 activity