Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-099 hydrochloride | 849335-07-5 | 99.9% | 301.10 g/mol | C9H9Cl2F3N4 | 10 MG
SDP

Catalog No. 5000286155
Encompass_Preferred

AC-099 hydrochloride is a research-grade small-molecule hydrochloride salt used in preclinical studies as a ligand for neuropeptide FF receptors. The compound is supplied as a solid and characterized by standard analytical data for laboratory use.

  • Research use only; not for human or veterinary use.
  • High reported purity, 99.9%.
  • Chemical formula C9H9Cl2F3N4 and molecular weight 301.10 g/mol.
  • Provided as a solid suitable for dissolution in common organic solvents.
  • Common applications include receptor pharmacology and in vivo preclinical assays.

Catalog No. 50-002-86155 Supplier Medchemexpress LLC Supplier No. HY154829A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AC-099 hydrochloride is a research-grade small-molecule hydrochloride salt used in preclinical studies as a ligand for neuropeptide FF receptors. The compound is supplied as a solid and characterized by standard analytical data for laboratory use.

  • Research use only; not for human or veterinary use.
  • High reported purity, 99.9%.
  • Chemical formula C9H9Cl2F3N4 and molecular weight 301.10 g/mol.
  • Provided as a solid suitable for dissolution in common organic solvents.
  • Common applications include receptor pharmacology and in vivo preclinical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.