Learn More
Medchemexpress LLC AC-099 (hydrochloride) | 849335-07-5 | C9H9Cl2F3N4 | 5 MG
SDP
Catalog No. 5000376283
Encompass_Preferred
AC-099 hydrochloride is a chemical compound primarily used in laboratory settings and for the manufacture of various substances. It is identified by its CAS number 849335-07-5 and has a chemical formula of C9H9Cl2F3N4.
  • Keep container tightly sealed in cool, well-ventilated area.
  • Store away from direct sunlight and sources of ignition.
  • Powder stability: 3 years at -20°C, 2 years at 4°C.
  • Stability in solvent: 6 months at -80°C.
  • Stable under recommended storage conditions.
Catalog No. 50-003-76283 Supplier Medchemexpress LLC Supplier No. HY154829A5MG
AC-099 hydrochloride is a chemical compound primarily used in laboratory settings and for the manufacture of various substances. It is identified by its CAS number 849335-07-5 and has a chemical formula of C9H9Cl2F3N4.
  • Keep container tightly sealed in cool, well-ventilated area.
  • Store away from direct sunlight and sources of ignition.
  • Powder stability: 3 years at -20°C, 2 years at 4°C.
  • Stability in solvent: 6 months at -80°C.
  • Stable under recommended storage conditions.