Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AC-099 hydrochloride | 849335-07-5 | C9H9Cl2F3N4 | 50 MG
SDP

Catalog No. 5000376488
Encompass_Preferred

AC-099 hydrochloride (compound 3) is a selective NPFF2R full agonist (EC50=1189 nM) and NPFF1R partial agonist (EC50=2370 nM). It has been shown to attenuate spinal nerve ligation-induced hypersensitivity in rats. This product is for research use only.

  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Appearance: Solid, white to off-white
  • Purity: 99.92%
  • Molecular weight: 301.10

Catalog No. 50-003-76488 Supplier Medchemexpress LLC Supplier No. HY154829A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AC-099 hydrochloride (compound 3) is a selective NPFF2R full agonist (EC50=1189 nM) and NPFF1R partial agonist (EC50=2370 nM). It has been shown to attenuate spinal nerve ligation-induced hypersensitivity in rats. This product is for research use only.

  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Appearance: Solid, white to off-white
  • Purity: 99.92%
  • Molecular weight: 301.10

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.