Learn More
Medchemexpress LLC AC-186 | 1421854-16-1 | 99.61% | 306.32 | 1 ML
SDP
Catalog No. 5000344017
Encompass_Preferred
AC-186 is a selective non-steroidal estrogen receptor β (ERβ) agonist with EC50 values of 6 nM for ERβ and 5000 nM for ERα. This compound has demonstrated gender-selective neuroprotective effects in a male rat model of Parkinson's disease by preventing motor, cognitive, and sensorimotor gating deficits and mitigating the loss of dopamine neurons in the substantia nigra. It also inhibits the growth of LNCaP cells.
  • Selective non-steroidal estrogen receptor β agonist
  • Demonstrates gender-selective neuroprotective effects
  • Inhibits growth of LNCaP cells in vitro
  • Suitable for research use only
Catalog No. 50-003-44017 Supplier Medchemexpress LLC Supplier No. HY11015710MM/1ML
AC-186 is a selective non-steroidal estrogen receptor β (ERβ) agonist with EC50 values of 6 nM for ERβ and 5000 nM for ERα. This compound has demonstrated gender-selective neuroprotective effects in a male rat model of Parkinson's disease by preventing motor, cognitive, and sensorimotor gating deficits and mitigating the loss of dopamine neurons in the substantia nigra. It also inhibits the growth of LNCaP cells.
  • Selective non-steroidal estrogen receptor β agonist
  • Demonstrates gender-selective neuroprotective effects
  • Inhibits growth of LNCaP cells in vitro
  • Suitable for research use only