Learn More
Medchemexpress LLC AC 187 | 151804-77-2 | 98.9% | 2890.21 | 10 MG
SDP
Catalog No. 5000422463
Encompass_Preferred
AC 187 is a potent and orally active amylin receptor antagonist. It demonstrates high selectivity for the amylin receptor compared to calcitonin and CGRP receptors. This compound exhibits neuroprotective effects by blocking amyloidβ protein (Aβ)-induced neurotoxicity and improving neuronal survival.
  • Potent and orally active amylin receptor antagonist
  • High selectivity for the amylin receptor
  • Exhibits neuroprotective effects
  • Blocks amyloidβ protein (Aβ)-induced neurotoxicity
  • Improves neuronal survival
  • Attenuates activation of caspases mediating Aβ-induced apoptotic cell death
  • Increases glucagon concentration
  • Accelerates gastric emptying of liquids
Catalog No. 50-004-22463 Supplier Medchemexpress LLC Supplier No. HYP139310MG
AC 187 is a potent and orally active amylin receptor antagonist. It demonstrates high selectivity for the amylin receptor compared to calcitonin and CGRP receptors. This compound exhibits neuroprotective effects by blocking amyloidβ protein (Aβ)-induced neurotoxicity and improving neuronal survival.
  • Potent and orally active amylin receptor antagonist
  • High selectivity for the amylin receptor
  • Exhibits neuroprotective effects
  • Blocks amyloidβ protein (Aβ)-induced neurotoxicity
  • Improves neuronal survival
  • Attenuates activation of caspases mediating Aβ-induced apoptotic cell death
  • Increases glucagon concentration
  • Accelerates gastric emptying of liquids