Learn More
Medchemexpress LLC AC 187 TFA, 10mg | Purity: 99.43% | Formula: C129H206F3N37O42
SDP
Catalog No. NC3662513
Encompass_Preferred
AC 187 TFA is a potent and orally active amylin receptor antagonist with an IC50 of 0.48 nM and a Ki of 0.275 nM. AC 187 TFA shows more selective for amylin receptor than calcitonin and CGRP receptors. AC 187 TFA has neuroprotective effects.
Catalog No. NC3662513 Supplier Medchemexpress LLC Supplier No. HYP1393A10MG
AC 187 TFA is a potent and orally active amylin receptor antagonist with an IC50 of 0.48 nM and a Ki of 0.275 nM. AC 187 TFA shows more selective for amylin receptor than calcitonin and CGRP receptors. AC 187 TFA has neuroprotective effects.