Learn More
Medchemexpress LLC Ac-261066 | 870773-76-5 | 99.3% | 353.41 | 50 MG
SDP
Catalog No. 5000346368
Encompass_Preferred
AC-261066 is a potent, orally available, and isoform-selective retinoic acid beta2 (RARbeta2) receptor agonist. It is intended for research use only and not for patient use.
  • Potent and orally available RARbeta2 receptor agonist
  • Isoform-selective for RARbeta2 receptor with a pEC50 of 8.0
  • Exhibits good oral bioavailability
Catalog No. 50-003-46368 Supplier Medchemexpress LLC Supplier No. HY10853210MM/1ML
AC-261066 is a potent, orally available, and isoform-selective retinoic acid beta2 (RARbeta2) receptor agonist. It is intended for research use only and not for patient use.
  • Potent and orally available RARbeta2 receptor agonist
  • Isoform-selective for RARbeta2 receptor with a pEC50 of 8.0
  • Exhibits good oral bioavailability