Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AC-263093 free base | 849459-86-5 | 99.8% | 319.98 | C8H8Br2N4 | 10MM 1ML
SDP

Catalog No. 5000237048
Encompass_Preferred

AC-263093 free base is a research-grade small molecule that functionally activates the neuropeptide FF receptor 2 (NPFFR2) and blocks NPFFR1; it is used in preclinical pharmacology studies related to opioid tolerance and receptor signaling. The compound is offered as a free base in solid form and as a ready-to-use 10 mM solution in DMSO for in vitro or in vivo preparation.

  • Functionally activates NPFFR2 and blocks NPFFR1.
  • Available as solid and as a 10 mM solution in DMSO.
  • High purity (99.8%).
  • Molecular formula C8H8Br2N4; molecular weight 319.98.
  • Recommended storage: powder at -20°C (long-term) or 4°C (short-term); in solvent at -80°C (long-term).
  • Intended for research use only.

Catalog No. 50-002-37048 Supplier Medchemexpress LLC Supplier No. HY12468010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AC-263093 free base is a research-grade small molecule that functionally activates the neuropeptide FF receptor 2 (NPFFR2) and blocks NPFFR1; it is used in preclinical pharmacology studies related to opioid tolerance and receptor signaling. The compound is offered as a free base in solid form and as a ready-to-use 10 mM solution in DMSO for in vitro or in vivo preparation.

  • Functionally activates NPFFR2 and blocks NPFFR1.
  • Available as solid and as a 10 mM solution in DMSO.
  • High purity (99.8%).
  • Molecular formula C8H8Br2N4; molecular weight 319.98.
  • Recommended storage: powder at -20°C (long-term) or 4°C (short-term); in solvent at -80°C (long-term).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.