Learn More
Medchemexpress LLC HY-124500 5mg , AC-4-130 CAS:1834571-82-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1245005MG
Encompass_Preferred
HY-124500 5mg AC-4-130 CAS: 1834571-82-2 AC-4-130 is a potent STAT5 SH2 domain inhibitor. AC-4-130 directly binds to STAT5 and disrupts STAT5 activation, dimerization, nuclear translocation, and STAT5-dependent gene transcription. AC-4-130 induces cell cycle arrest and apoptosis in FLT3-ITD-driven leukemic cells. AC-4-130 has anti-cancer activity and can efficiently block pathological levels of STAT5 activity in acute myeloid leukemia (AML). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9235