Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AC-42 hydrochloride | 447407-36-5 | 98.4% | 337.93 g·mol⁻¹ | C20H32ClNO | 10 MG
SDP

Catalog No. 5000308065
Encompass_Preferred

AC-42 hydrochloride is the hydrochloride salt of AC-42, an allosteric agonist selective for the muscarinic M1 receptor used for in vitro receptor pharmacology and signaling studies.

  • Allosteric agonist of muscarinic M1 receptor; stimulates inositol phosphate accumulation and calcium mobilization in CHO cells.
  • CAS 447407-36-5.
  • Molecular formula C20H32ClNO; molecular weight 337.93 g·mol⁻¹.
  • Purity 98.42% (as supplied).
  • Appearance off-white to light brown solid; store sealed and away from moisture at 4°C; in solvent store at -80°C for up to 6 months or -20°C for 1 month.
  • Available in small research quantities, including 10 MG packaging.

Catalog No. 50-003-08065 Supplier Medchemexpress LLC Supplier No. HY118806A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AC-42 hydrochloride is the hydrochloride salt of AC-42, an allosteric agonist selective for the muscarinic M1 receptor used for in vitro receptor pharmacology and signaling studies.

  • Allosteric agonist of muscarinic M1 receptor; stimulates inositol phosphate accumulation and calcium mobilization in CHO cells.
  • CAS 447407-36-5.
  • Molecular formula C20H32ClNO; molecular weight 337.93 g·mol⁻¹.
  • Purity 98.42% (as supplied).
  • Appearance off-white to light brown solid; store sealed and away from moisture at 4°C; in solvent store at -80°C for up to 6 months or -20°C for 1 month.
  • Available in small research quantities, including 10 MG packaging.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.