Learn More
Enzo Life Sciences AC-55469 (10mg). CAS: 59662-49-6
SDP
Supplier:  Enzo Life Sciences BMLGR1120010
Encompass_Preferred
AC-55649 is potent and subtype selective agonist at RARβ2 ( EC50=6.9 µM). It inhibits proliferation of MCF-7 cells and induces differentiation in a neuronal cell line. Purity: ≥98% (HPLC). Long Term Storage: Ambient.
Catalog No. 50-201-0925