Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AC-55541 | 916170-19-9 | 99.4% | C25H20BrN5O3 | 100 MG
SDP

Catalog No. 5000354254
Encompass_Preferred

AC-55541 is a highly selective agonist for protease-activated receptor 2 (PAR2), demonstrating a pEC50 of 6.7. It does not show activity at other PAR subtypes or at over 30 additional receptors associated with nociception and inflammation. AC-55541 has pEC50 values of 5.9 and 6.6 in PI hydrolysis assays and Ca2+ mobilization assays, and exhibits pronociceptive activity in vivo.

  • Highly selective agonist for PAR2 (pEC50=6.7)
  • No activity at other PAR subtypes or over 30 other receptors involved in nociception and inflammation
  • Purity 99.4%
  • Molecular weight 518.36
  • Appears as a white to off-white solid
  • Soluble in DMSO at ≥ 51 mg/mL
  • Suppresses poly I:C-induced CXCL10 mRNA in NHBE cells in vitro
  • Exhibits pronociceptive activity in vivo

Catalog No. 50-003-54254 Supplier Medchemexpress LLC Supplier No. HY14350100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AC-55541 is a highly selective agonist for protease-activated receptor 2 (PAR2), demonstrating a pEC50 of 6.7. It does not show activity at other PAR subtypes or at over 30 additional receptors associated with nociception and inflammation. AC-55541 has pEC50 values of 5.9 and 6.6 in PI hydrolysis assays and Ca2+ mobilization assays, and exhibits pronociceptive activity in vivo.

  • Highly selective agonist for PAR2 (pEC50=6.7)
  • No activity at other PAR subtypes or over 30 other receptors involved in nociception and inflammation
  • Purity 99.4%
  • Molecular weight 518.36
  • Appears as a white to off-white solid
  • Soluble in DMSO at ≥ 51 mg/mL
  • Suppresses poly I:C-induced CXCL10 mRNA in NHBE cells in vitro
  • Exhibits pronociceptive activity in vivo

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.