Learn More
Medchemexpress LLC HY-108526 10mg Medchemexpress, AC-55649 CAS:59662-49-6 Purity:>98%
SDP
Catalog No. 501968570
Encompass_Preferred
Medchemexpress, HY-108526 10mg AC-55649 CAS:59662-49-6 AC-55649 is a potent, highly isoform-selective agonist of human RARβ2 receptor, with a pEC50 of 6.9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8570 Supplier Medchemexpress LLC Supplier No. HY10852610MG
Medchemexpress, HY-108526 10mg AC-55649 CAS:59662-49-6 AC-55649 is a potent, highly isoform-selective agonist of human RARβ2 receptor, with a pEC50 of 6.9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.