Learn More
Bachem Ac-(6-O-stearoyl)-muramyl-Ala-D-Glu-NH2
SDP
Supplier:  Bachem 4015905.0001
Encompass_Preferred
Ac-(6-O-stearoyl)-muramyl-Ala-D-Glu-NH2, 60398-08-5, C37H66N4O12, Mr 758.95, 1mg. Ac-(6-O-stearoyl)-muramyl-Ala-D-Isogln-OH, L18-MDP. The lipidated L18 MDP showed a higher activity than unmodified MDP. L18 MDP acts as a NOD2 receptor agonist.
Catalog No. 50-259-252