Learn More
TARGETMOL CHEMICALS INC AC-73 10MG
SDP
Catalog No. 502879605
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. AC-73 is an orally available Cluster of Differentiation 147 (CD147) inhibitor with high bioavailability that selectively disrupts the dimerization of CD147 (the binding site is in the N-terminal IgC2 domain of CD147 including Glu64 and Glu73), resulting in inhibition of the CD147/ERK1/2/STAT3/MMP-2 pathway and inhibition of liver cancer cell motility and invasion. AC-73 has antiproliferative activity and induces autophagy in leukemia cells. Purity 98.95%
Catalog No. 50-287-9605 Supplier TARGETMOL CHEMICALS INC Supplier No. T1409110MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. AC-73 is an orally available Cluster of Differentiation 147 (CD147) inhibitor with high bioavailability that selectively disrupts the dimerization of CD147 (the binding site is in the N-terminal IgC2 domain of CD147 including Glu64 and Glu73), resulting in inhibition of the CD147/ERK1/2/STAT3/MMP-2 pathway and inhibition of liver cancer cell motility and invasion. AC-73 has antiproliferative activity and induces autophagy in leukemia cells. Purity 98.95%