Learn More
TARGETMOL CHEMICALS INC AC 7739 1MG
SDP
Catalog No. 502877909
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AC 7739 is a microtubule protein binding agent with anticancer and antitumor activity that inhibits advanced solid tumors and in situ transplanted tumors. Purity 99.38%
Catalog No. 50-287-7909 Supplier TARGETMOL CHEMICALS INC Supplier No. T71924L1MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AC 7739 is a microtubule protein binding agent with anticancer and antitumor activity that inhibits advanced solid tumors and in situ transplanted tumors. Purity 99.38%