Learn More
Enzo Life Sciences Ac-AAVALLPAVLLALLAPDEVD-CHO (1mg)
SDP
Catalog No. 502008258
Encompass_Preferred
Caspase inhibitor. Reversible, cell permeable inhibitor of caspase-3. Also inhibits caspase-6, -7, -8 and -10. Alternative name: Caspase 3 inhibitor (aldehyde). Formula: C94H158N20O27. MW: 2000.4. Purity: ≥95% (HPLC). Appearance: White to off-white powder. Formulation: Lyophilized. TFA salt. Peptide content: 65-95%. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: After dissolving, prepare aliquots and store at -20°C. Keep cool and dry.
Catalog No. 50-200-8258 Supplier Enzo Life Sciences Supplier No. ALX260046M001
Caspase inhibitor. Reversible, cell permeable inhibitor of caspase-3. Also inhibits caspase-6, -7, -8 and -10. Alternative name: Caspase 3 inhibitor (aldehyde). Formula: C94H158N20O27. MW: 2000.4. Purity: ≥95% (HPLC). Appearance: White to off-white powder. Formulation: Lyophilized. TFA salt. Peptide content: 65-95%. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: After dissolving, prepare aliquots and store at -20°C. Keep cool and dry.