Learn More
Cayman Chemical Ac-AAVALLPAVLLALLAP-IETD-CHO (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502940197
Encompass_Preferred
A composite of a caspase-8 inhibitor and a cell-permeable hydrophobic sequence; an inhibitor of caspase-8 and granzyme B; prevents death of SKOV3 ovarian carcinoma cells transfected with SAGA at 20 µM
Catalog No. 50-294-0197 Supplier Cayman Chemical Supplier No. 274381 MG
A composite of a caspase-8 inhibitor and a cell-permeable hydrophobic sequence; an inhibitor of caspase-8 and granzyme B; prevents death of SKOV3 ovarian carcinoma cells transfected with SAGA at 20 µM