Learn More
Cayman Chemical Ac-AAVALLPAVLLALLAP-VAD-CHO (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502941120
Encompass_Preferred
A composite of a non-selective caspase inhibitor and a cell-permeable hydrophobic sequence; a non-selective caspase inhibitor
Catalog No. 50-294-1120 Supplier Cayman Chemical Supplier No. 274251 MG
A composite of a non-selective caspase inhibitor and a cell-permeable hydrophobic sequence; a non-selective caspase inhibitor