Learn More
Cayman Chemical Ac-AAVAlPAVlAlAP-VAD-CHO 500ug
SDP
Catalog No. 502939163
Encompass_Preferred
A composite of a non-selective caspase inhibitor and a cell-permeable hydrophobic sequence; a non-selective caspase inhibitor
Catalog No. 50-293-9163 Supplier Cayman Chemical Supplier No. 27425500 UG
A composite of a non-selective caspase inhibitor and a cell-permeable hydrophobic sequence; a non-selective caspase inhibitor