Learn More
Ubpbio A fluorogenic substrate of proteasomes, preferentially cleaved by immunoproteasomes.
Catalog No. 502339249
Encompass_Preferred
Ac-ANW-AMC is a fluorogenic substrate of the chymotrypsin-like activity of proteasomes. This substrate is preferentially cleaved by immunoproteasomes compared to constitutive proteasomes. The AMC fluorescence can be detected by a fluorimeter or a plate reader using excitation/emission wavelengths at 360 nm/460 nm, respectively. This substrate can be used to determine the chymotrypsin-like activity of immunoproteasomes. Working concentration is 50 - 200 µM.
Catalog No. 50-233-9249 Supplier Ubpbio Supplier No. G1121
Ac-ANW-AMC is a fluorogenic substrate of the chymotrypsin-like activity of proteasomes. This substrate is preferentially cleaved by immunoproteasomes compared to constitutive proteasomes. The AMC fluorescence can be detected by a fluorimeter or a plate reader using excitation/emission wavelengths at 360 nm/460 nm, respectively. This substrate can be used to determine the chymotrypsin-like activity of immunoproteasomes. Working concentration is 50 - 200 µM.