Learn More
American Radiolabeled Chemicals Inc AC ACID-1-14C NA SALT A 1LC
SDP
Catalog No. 5090105286
Encompass_Preferred
Acetic acid-1-14C sodium salt 1mCi in ethanol, 1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 82.0
Catalog No. 50-901-05286 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 0101A1MC
Acetic acid-1-14C sodium salt 1mCi in ethanol, 1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 82.0