Learn More
Vernier™ Vernier AC Adapter
SDPSCI_ED
Catalog No. S15990ND Shop All Vernier Software Products
Change view
Click to view available options
Voltage:
6 V
Catalog No. Voltage
S15990ND 6 V
1 options
Catalog No. S15990ND Supplier Vernier™ Supplier No. IPS
Add to Cart
Add to Cart
Includes
Vernier AC Adapter
This is a power supply (6 volt, DC, tip-negative) connects to your LabPro, Wireless Dynamics Sensor System, Digital Control Unit, CBL 2, or CBL to AC power.
  • This is a 6-volt, DC, tip-negative power supply for use with Vernier™ LabPro, CBL 2, Digital Control Unit, Stir Station and Wireless Dynamics Sensor System.
  • If you need an international power supply for 220-volts, use International Power Supply for LabPro, WDSS, and DCU.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Equipment) Digital Control Unit, Wireless Dynamics Sensor System, CBL 2, LabPro, Stir Station
Voltage 6 V
For Use With (Application) Educational use only
Power Supply Details DC Tip-Negative
Warranty 5-year limited warranty