Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Universal AC Adapter

Catalog No. 14380974 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With (Equipment):
HandyStep Touch / Touch S
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-380-974 HandyStep Touch / Touch S
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-380-974 Supplier BRANDTECH™ BRAND™ Supplier No. 705250
Add to Cart
Add to Cart

For use with HandyStep™ touch and HandyStep™ touch S.

  • Includes USB cable and 5 country adapters
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, C/US
Autoclavable Not Autoclavable
Color Black
For Use With BRAND HandyStep Touch/Touch S
Material Plastic
Type AC Adapter/Power Supply
For Use With (Equipment) HandyStep Touch / Touch S
Plug Type US, UK, CEE, CN
Quantity 1
Electrical Requirements 100 to 240 V, 50/60 Hz
Amperage 0.5 A
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.