Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ AC Adapter for M210 Handheld Label Maker

Catalog No. 19000748 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-000-748 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-000-748 Supplier Brady™ Supplier No. M210AC
Add to Cart
Add to Cart
This item is not returnable. View return policy

Brady™ AC Adaptor allows you to power your M210 Label Printer without the need for recharging or replacing batteries.

  • This AC Adaptor allows you to power your M210 Label Printer without the need for recharging or replacing batteries.
  • Package includes an AC adapter/charger (printer and battery pack sold separately).
  • Power supply voltage: 110 V.
  • Power supply formatted for North America.

Specifications

Product Type Printer
Color Black
Display Type None
For Use With (Equipment) Brady Labels, Brady M210-LAB Label Printer, BMP21 LAB Label Printer, BMP21-PLUS Label Printer, BMP21 Label Printer
Material Plastic
Width (English) 6.9 in.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.