Learn More
Brady AC Adapter for Portable Printers
Catalog No. 17467584 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
BMP41 Label Printer, Brady M410 Label Printer, BMP51 Label Printer, BMP53 Label Printer, Brady M510 Label Printer, BMP61 Labeling System, BradyPrinter M610, BradyPrinter M611
Catalog No. For Use With (Equipment)
17-467-584 BMP41 Label Printer, Brady M410 Label Printer, BMP51 Label Printer, BMP53 Label Printer, Brady M510 Label Printer, BMP61 Labeling System, BradyPrinter M610, BradyPrinter M611
1 options
Catalog No. 17-467-584 Supplier Brady Supplier No. M415161AC
Add to Cart
Add to Cart
Work the entire day using this AC Adapter/Battery Charger. It powers the M410, M510, M610 and M611 label printers and charges the related battery packs.
  • Work the entire day using this AC Adapter/Battery Charger.
  • It powers the M410, M510, M610 and M611 label printers and charges the related battery packs.
  • Use this adapter to power and charge your M410, M510, M610 and M611 label printers.
  • Includes an AC Adapter/Charger.
  • Power supply voltage: 110 - 240 V.
  • Power supply formatted for North America.
Specifications
Product Type AC Adapter
For Use With (Equipment) BMP41 Label Printer, Brady M410 Label Printer, BMP51 Label Printer, BMP53 Label Printer, Brady M510 Label Printer, BMP61 Labeling System, BradyPrinter M610, BradyPrinter M611
Voltage 110/240 V
WARNING: Reproductive Harm - www.P65Warnings.ca.gov