Learn More
Description
- Use this adapter to power and charge your M610 and M611 label printers
- Includes an AC Adapter/Charger
- Power supply voltage: 110/240 V
- Power supply formatted for North America
Specifications
Specifications
| Product Type | AC Adapter |
| Color | Black |
| For Use With (Equipment) | BradyPrinter M611, BradyPrinter M610, BMP41 Label Printer, BMP51 Label Printer, BMP53 Label Printer, BMP61 Labeling System |
| Adhesive Type | Non-adhesive |
| Autoclavable | Not Autoclavable |
| Sterility | Non-sterile |
| Includes | AC Adapter, Battery Charger |
| Voltage | 110/240 V |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.