Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-Ala-OH-d3 | 1485548-36-4 | 134.15 | 1 MG
SDP

Catalog No. 5000445732
Encompass_Preferred

Ac-Ala-OH-d3 is a chemical compound with the molecular formula C5H6D3NO3 and a molecular weight of 134.15. It has a high purity of 99.0% and an isotopic enrichment of 99.8%. This product appears as a white to off-white solid.

  • Molecular formula: C5H6D3NO3
  • Molecular weight: 134.15
  • Purity: 99.0%
  • Isotopic enrichment: 99.8%
  • Appearance: White to off-white solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-45732 Supplier Medchemexpress LLC Supplier No. HYW004066S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-Ala-OH-d3 is a chemical compound with the molecular formula C5H6D3NO3 and a molecular weight of 134.15. It has a high purity of 99.0% and an isotopic enrichment of 99.8%. This product appears as a white to off-white solid.

  • Molecular formula: C5H6D3NO3
  • Molecular weight: 134.15
  • Purity: 99.0%
  • Isotopic enrichment: 99.8%
  • Appearance: White to off-white solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.