Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-Ala-OH-d3 | 1485548-36-4 | 134.15 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYW004066S5MG

Encompass_Preferred

Ac-Ala-OH-d3 is presented as a white to off-white solid. This product has an isotopic enrichment of 99.8% and a purity of 99.0%, and is suitable for various research applications. It complies with given specifications.

  • Appearance: White to off-white solid
  • Purity: 99.0%
  • Isotopic enrichment: 99.8%
  • Tested and complies with specifications
  • For research use only

Catalog No. 50-004-47145


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.