Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-β-Ala-OH (N-Acetyl-β-alanine) | 3025-95-4 | 98.0% | C5H9NO3 | 50 G
SDP

Catalog No. 5000434989
Encompass_Preferred

Ac-β-Ala-OH (N-Acetyl-β-alanine) is an abnormal amino acid metabolite that functions as a mono-N-protected amino acid (MPAA) ligand. It is supplied as a white to off-white solid with a purity of 98.0%.

  • Molecular weight: 131.13
  • Chemical formula: C5H9NO3
  • CAS number: 3025-95-4
  • Appearance: White to off-white solid
  • Purity: 98.0%
  • Structure classification: Amino acids; endogenous metabolite
  • Shipping: Room temperature in continental US
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • In vitro solubility: 100 mg/mL in DMSO
  • For research use only

Catalog No. 50-004-34989 Supplier Medchemexpress LLC Supplier No. HY6600750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-β-Ala-OH (N-Acetyl-β-alanine) is an abnormal amino acid metabolite that functions as a mono-N-protected amino acid (MPAA) ligand. It is supplied as a white to off-white solid with a purity of 98.0%.

  • Molecular weight: 131.13
  • Chemical formula: C5H9NO3
  • CAS number: 3025-95-4
  • Appearance: White to off-white solid
  • Purity: 98.0%
  • Structure classification: Amino acids; endogenous metabolite
  • Shipping: Room temperature in continental US
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • In vitro solubility: 100 mg/mL in DMSO
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.