Learn More
Bachem Ac-Ala-pNA
SDP
Supplier:  Bachem 4014152.0250
Encompass_Preferred
Ac-Ala-pNA, 35978-75-7, C11H13N3O4, Mr 251.24, 250mg. AANA, substrate for acylaminoacyl peptidases (APH, acyl peptide hydrolase), an enzyme able to degrade monomeric and oligomeric amyloid peptides.
Catalog No. 50-260-420