Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-ANW-AMC | 2357123-49-8 | 99.4% | 10 MG
SDP

Catalog No. 5000297820
Encompass_Preferred

Ac-ANW-AMC is a fluorogenic peptide substrate used to measure immunoproteasome β5i activity in vitro. Proteolytic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal suitable for kinetic or end-point assays.

  • releases AMC fluorophore upon proteolytic cleavage for fluorescence detection.
  • reported excitation/emission: 380 nm/460 nm; assay notes recommend 345 nm/445 nm for β5i measurements.
  • high purity (99.4%) for reliable biochemical assays.
  • molecular weight 588.61 g/mol; formula C30H32N6O7.
  • available in small quantities (for example, 10 mg) for research use.
  • store at -20°C and protect from light; in solution, store at -80°C for long-term stability.

Catalog No. 50-002-97820 Supplier Medchemexpress LLC Supplier No. HYD170510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-ANW-AMC is a fluorogenic peptide substrate used to measure immunoproteasome β5i activity in vitro. Proteolytic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal suitable for kinetic or end-point assays.

  • releases AMC fluorophore upon proteolytic cleavage for fluorescence detection.
  • reported excitation/emission: 380 nm/460 nm; assay notes recommend 345 nm/445 nm for β5i measurements.
  • high purity (99.4%) for reliable biochemical assays.
  • molecular weight 588.61 g/mol; formula C30H32N6O7.
  • available in small quantities (for example, 10 mg) for research use.
  • store at -20°C and protect from light; in solution, store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.