Learn More
Bachem Ac-Arg-Cys-Met-5-aminopentanoyl-Arg-Val-Tyr-5-aminopentanoyl-Cys-NH2
SDP
Supplier:  Bachem 4045499.0001
Encompass_Preferred
Ac-Arg-Cys-Met-5-aminopentanoyl-Arg-Val-Tyr-5-aminopentanoyl-Cys-NH2 Trifluoroacetate, 353487-64-6, C49H82N16O11S3, Mr 1167.49, 1mg. Ac-Arg-Cys-Met-Ava-Arg-Val-Tyr-Ava-Cys-NH2. Full competitive MCH-1 receptor antagonist devoid of agonist potency for the human MCH-1 receptor even at micromolar concentrations (Kb = 3.6 nM).
Catalog No. 50-383-255