Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-Arg-Gly-Lys(Ac)-AMC | 660846-97-9 | 99.9% | 600.67 | 1 MG
SDP

Catalog No. 5000424223
Encompass_Preferred

Ac-Arg-Gly-Lys(Ac)-AMC is a peptide substrate for Histone Deacetylases (HDACs). It is intended for research purposes only. This compound facilitates in vitro studies where, after initial HDAC inhibition, the substrate is introduced to reaction tubes, and the enzymatic activity is measured before being terminated by a stop solution.

  • Substrate for HDAC studies.
  • High purity of 99.9%.
  • Appears as a white to off-white solid.
  • Soluble in H2O at concentrations of 50 mg/mL or greater.
  • Store sealed, away from moisture and light, under nitrogen.
  • Powder stability: 2 years at -80°C, 1 year at -20°C.
  • Solution stability: 6 months at -80°C, 1 month at -20°C.

Catalog No. 50-004-24223 Supplier Medchemexpress LLC Supplier No. HYP24621MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-Arg-Gly-Lys(Ac)-AMC is a peptide substrate for Histone Deacetylases (HDACs). It is intended for research purposes only. This compound facilitates in vitro studies where, after initial HDAC inhibition, the substrate is introduced to reaction tubes, and the enzymatic activity is measured before being terminated by a stop solution.

  • Substrate for HDAC studies.
  • High purity of 99.9%.
  • Appears as a white to off-white solid.
  • Soluble in H2O at concentrations of 50 mg/mL or greater.
  • Store sealed, away from moisture and light, under nitrogen.
  • Powder stability: 2 years at -80°C, 1 year at -20°C.
  • Solution stability: 6 months at -80°C, 1 month at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.