Learn More
AdipoGen Ac-Arg-Leu-Arg-AMC (5 mg)
SDP
Catalog No. 501490031
Encompass_Preferred
Ac-Arg-Leu-Arg-AMC (5 mg), AdipoGen Life Sciences. Chemical. CAS: 929903-87-7 (free base). Formula: C30H46N10O6 . C2HF3O2. MW: 642.8 . 114.0. Fluorogenic tri-peptide substrate for measuring the trypsin-like peptidase activity of the 20S proteasome. It is cleaved exclusively at the amide R-AMC bond to release fluorescent product. Shows low Km and high specific activity. Excitation: 380nm. Emission: 460nm. This substrate is useful for inhibitor screening and kinetic analysis.
Catalog No. 50-149-0031 Supplier AdipoGen Supplier No. AGCP30013M005
Ac-Arg-Leu-Arg-AMC (5 mg), AdipoGen Life Sciences. Chemical. CAS: 929903-87-7 (free base). Formula: C30H46N10O6 . C2HF3O2. MW: 642.8 . 114.0. Fluorogenic tri-peptide substrate for measuring the trypsin-like peptidase activity of the 20S proteasome. It is cleaved exclusively at the amide R-AMC bond to release fluorescent product. Shows low Km and high specific activity. Excitation: 380nm. Emission: 460nm. This substrate is useful for inhibitor screening and kinetic analysis.