Learn More
AdipoGen Ac-Arg-Leu-Arg-AMC
SDP
Catalog No. 502562450
Encompass_Preferred
Chemical. CAS 929903-87-7 free base. Formula C30H46N10O6 . C2HF3O2. MW 642.8 . 114.0. Fluorogenic tri-peptide substrate for measuring the trypsin-like peptidase activity of the 20S proteasome. It is cleaved exclusively at the amide R-AMC bond to release fluorescent product. Shows low Km and high specific activity. Excitation 380nm. Emission 460nm. This substrate is useful for inhibitor screening and kinetic analysis.
Catalog No. 50-256-2450 Supplier AdipoGen Supplier No. AGCP30013M001
Chemical. CAS 929903-87-7 free base. Formula C30H46N10O6 . C2HF3O2. MW 642.8 . 114.0. Fluorogenic tri-peptide substrate for measuring the trypsin-like peptidase activity of the 20S proteasome. It is cleaved exclusively at the amide R-AMC bond to release fluorescent product. Shows low Km and high specific activity. Excitation 380nm. Emission 460nm. This substrate is useful for inhibitor screening and kinetic analysis.