Learn More
Enzo Life Sciences Ac-Arg-Leu-Arg-AMC (5mg)
SDP
Catalog No. 502010335
Encompass_Preferred
This fluorogenic peptide is often the substrate of choice for assaying the trypsin-like activity of purified proteasomes. Benefits include a low Km, high specific activity, and the fact that the substrate is cleaved exclusively at the R-AMC bond. Alternative name: Ac-RLR-AMC. Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO, methanol or aqueous solutions. Long Term Storage: -20°C.
Catalog No. 50-201-0335 Supplier Enzo Life Sciences Supplier No. BMLAW97850005
Add to Cart
Add to Cart
This fluorogenic peptide is often the substrate of choice for assaying the trypsin-like activity of purified proteasomes. Benefits include a low Km, high specific activity, and the fact that the substrate is cleaved exclusively at the R-AMC bond. Alternative name: Ac-RLR-AMC. Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO, methanol or aqueous solutions. Long Term Storage: -20°C.