Learn More
Ac-Asp-Glu-Val-Asp-7-Amino-4-TriFluoromethylcoumarin, MP Biomedicals™
Catalog No. 03AFC13825 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03AFC13825 25 mg
03AFC13805 5 mg
03AFC13810 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03AFC13825 Supplier MP Biomedicals, Inc Supplier No. 03AFC13825
Add to Cart
Add to Cart
Ac-Asp-Glu-Val-Asp-7-Amino-4-TriFluoromethylcoumarin is a fluorescent substrate.
  • Ac-DEVD-AFC is a sensitive fluorescent substrate for the assay of caspase-3 and CPP-32 activity.
  • The trifluoromethyl substituent of the fluorophore improves the membrane permeability of the substrate.
  • Protein or Enzyme Type: Activators, Inhibitors and Substrates
TRUSTED_SUSTAINABILITY
Specifications
Form Lyophilized
Quantity 25 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.