Learn More
Bachem Ac-Asp-Glu-Val-Asp-AMC
SDP
Catalog No. 50260338
Encompass_Preferred
Ac-Asp-Glu-Val-Asp-AMC Ammonium, 169332-61-0, C30H37N5O13, Mr 675.65, 5mg. Ac-DEVD-AMC, Apopain Substrate. A continuous fluorometric assay employing Ac-DEVD-AMC has been developed for the measurement of caspase-3, a cysteine protease also called apopain, Yama or CPP-32. The design of this substrate Ac-DEVD-AMC was based on the substrate motif that has been used successfully with caspase-1, using the poly(ADP-ribose) polymerase cleavage site P1-P4 tetrapeptide.
Catalog No. 50-260-338 Supplier Bachem Supplier No. 4026262.0005
Ac-Asp-Glu-Val-Asp-AMC Ammonium, 169332-61-0, C30H37N5O13, Mr 675.65, 5mg. Ac-DEVD-AMC, Apopain Substrate. A continuous fluorometric assay employing Ac-DEVD-AMC has been developed for the measurement of caspase-3, a cysteine protease also called apopain, Yama or CPP-32. The design of this substrate Ac-DEVD-AMC was based on the substrate motif that has been used successfully with caspase-1, using the poly(ADP-ribose) polymerase cleavage site P1-P4 tetrapeptide.