Learn More
Carbosynth LLC. Ac-Asp-Glu-Val-Asp-MCA [Ac-DEVD-MCA] (5 mg vial), 1ea, MW: 675.64, Molecular formula: C30H37N5O13, CAS# 169332-61-0
SDP
Supplier:  Carbosynth LLC. SAP3171V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Acetyl-L-Aspartyl-L-Glutamyl-L-Valyl-L-Aspartic Acid -(4-Methyl-Coumaryl-7-Amide), Ac-Asp-Glu-Val-Asp-AMC, Ac-DEVD-MCA, Ac-DEVD-AMC, Purity: , Grade A, Application: Substrate for Caspase-3/7/8, Storage Temperature: -20C, References: D.W. Nicholson, A. Ali, N.A. Thornberry, J.P. Vaillancourt, C.K. Ding, M. Gallant, Y. Gareau, P.R. Griffin, M. Labelle, Y.A. Lazebnik, N.A. Munday, S.M. Raju, M.E. Smulson, T.-T. Yamin, V.L. Yu, and D.K. Miller, Nature, 376, 37 (1995)., N.A. Thornberry, T.A. Rano, E.P. Peterson, D.M. Rasper, T. Timkey, M. Garcia-Calvo, V.M. Houtzager, P.A. Nordstrom, S. Roy, J.P. Vaillancourt, K.T. Chapman, and D.W. Nicholson, J. Biol. Chem., 272, 17907 (1997). Grade A
Catalog No. 50-168-7028