Learn More
MSC Ebm-Papst Blower, 115 VAC, 152 CFM, Centrifugal, 1 ea
Catalog No. 5000507150
Encompass
This centrifugal blower is designed for efficient air movement in thermal management and HVAC applications. It features a robust construction with a die-cast aluminum housing and a steel impeller, operating on a 115 VAC power supply to deliver an airflow of 152 CFM.
  • Features ball bearing design for reliable operation
  • Equipped with an internal thermal overload protector
  • Offers IP44 degree of protection depending on installation
  • Includes 4 wire leads for termination
  • Operates at a nominal frequency of 60 Hz
  • Constructed with materials compliant with RoHS3 standards
Catalog No. 50-005-07150 Supplier MSC Supplier No. 76940071
This centrifugal blower is designed for efficient air movement in thermal management and HVAC applications. It features a robust construction with a die-cast aluminum housing and a steel impeller, operating on a 115 VAC power supply to deliver an airflow of 152 CFM.
  • Features ball bearing design for reliable operation
  • Equipped with an internal thermal overload protector
  • Offers IP44 degree of protection depending on installation
  • Includes 4 wire leads for termination
  • Operates at a nominal frequency of 60 Hz
  • Constructed with materials compliant with RoHS3 standards