Learn More
Medchemexpress LLC Ac-CoA Synthase Inhi 25mg | 508186-14-9 | 25MG
SDP
Catalog No. 5000241858
Encompass_Preferred
Ac-CoA Synthase Inhibitor1 is a potent reversible acetate-dependent acetyl-CoA synthetase 2 (ACSS2) inhibitor with an IC50 of 0 6 M[1 Ac-CoA Synthase Inhibitor1 inhibits the respiratory syncytial virus (RSV)[2
Catalog No. 50-002-41858 Supplier Medchemexpress LLC Supplier No. HY10403225MG
Ac-CoA Synthase Inhibitor1 is a potent reversible acetate-dependent acetyl-CoA synthetase 2 (ACSS2) inhibitor with an IC50 of 0 6 M[1 Ac-CoA Synthase Inhibitor1 inhibits the respiratory syncytial virus (RSV)[2