Learn More
Medchemexpress LLC Ac-CoA synthase inhibitor 1 | 508186-14-9 | 99.7% | 410.51 g/mol | C20H18N4O2S2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10403210MM 1ML
Encompass_Preferred
Ac-CoA synthase inhibitor 1 is a small-molecule inhibitor of acetyl-CoA synthetase 2 (ACSS2) for research applications. It is supplied as a 10 mM solution in DMSO and is also available as a powder; characterized by CAS 508186-14-9, molecular formula C20H18N4O2S2, and molecular weight 410.51 g/mol.
  • Targets acetyl-CoA synthetase 2 (ACSS2) for pathway and antiviral research.
  • Supplied as a 10 mM solution in DMSO, ready for in vitro use.
  • High purity: 99.7%.
  • Light yellow solid appearance when dry.
  • Stable under recommended storage: powder -20°C or 4°C; in solvent -80°C or -20°C.
  • Structure and identifiers provided for reproducibility.
Catalog No. 50-002-42554