Learn More
CPC Scientific Ac-Cys-Glu-His-DPhe-Arg-Trp-Cys-Lys-Pro-Val-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific MSHS005A
Encompass
A superagonist with ultraprolonged melanotropic activity in frog and lizard skin bioassays. It exhibits a complete resistance to tryptic degradation. ONE-LETTER SEQUENCE: Ac-CEH-DPhe-RWCKPV-NH2 (Cys1 and 7 bridge)MOLECULAR FORMULA: C61H86N18O13S2MOLECULAR WEIGHT:1343.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [91050-39-4], SYNONYMS: melanotropinsRESEARCH AREA: HormonalREFERENCES: W.L. Cody et al., J. Med. Chem., 28, 683 (1985)
Catalog No. 50-210-9841